Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764175.1 | XP_008762397.1 | 3 - 4 | 769 | 72 | |
XM_008764176.1 | XP_008762398.1 | 3 - 4 | 887 | 72 | ||
XM_008764177.1 | XP_008762399.1 | 4 - 5 | 1863 | 72 | ||
XM_008764178.1 | XP_008762400.1 | 3 - 4 | 718 | 72 | ||
XM_008764179.1 | XP_008762401.1 | 3 - 4 | 1689 | 72 | ||
XM_575911.5 | XP_575911.4 | 4 - 5 | 1446 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |