Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019236.1 | NP_062109.1 | 1 - 2 | 167 | 79 | |
GenBank mRNA | D14029.1 | - | 1 - 2 | 167 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |