Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sesn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109358.2 | NP_001102828.1 | 6 - 7 | 1203 | 128 | |
GenBank mRNA | BC166423.1 | - | 6 - 7 | 1203 | 7096599 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |