Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007554.1 | NP_001007555.1 | 5 - 6 | 656 | 73 | |
XM_008764273.1 | XP_008762495.1 | 5 - 6 | 1423 | 73 | ||
XM_008764274.1 | XP_008762496.1 | 4 - 5 | 586 | 73 | ||
XM_008764275.1 | XP_008762497.1 | 5 - 6 | 655 | 73 | ||
XM_008764276.1 | XP_008762498.1 | 5 - 6 | 806 | 73 | ||
GenBank mRNA | BC086398.1 | - | 5 - 6 | 656 | 7303205 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |