Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109438.1 | NP_001102908.1 | 2 - 3 | 646 | 63 | |
XM_006238901.2 | XP_006238963.1 | 2 - 3 | 626 | 63 | ||
XM_006238902.2 | XP_006238964.1 | 2 - 3 | 626 | 63 | ||
XM_006238903.2 | XP_006238965.1 | 2 - 3 | 626 | 63 | ||
XM_006238904.2 | XP_006238966.1 | 2 - 3 | 626 | 63 | ||
XM_006238906.2 | XP_006238968.1 | 2 - 3 | 626 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |