Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191871.1 | NP_001178800.1 | 12 - 13 | 2354 | 65 | |
XM_006239232.2 | XP_006239294.1 | 12 - 13 | 2392 | 65 | ||
XM_006239233.2 | XP_006239295.1 | 12 - 13 | 2392 | 65 | ||
XM_008764263.1 | XP_008762485.1 | 12 - 13 | 2393 | 65 | ||
XM_008764264.1 | XP_008762486.1 | 12 - 13 | 7951 | 65 | ||
XM_008764265.1 | XP_008762487.1 | 13 - 14 | 3094 | 65 | ||
XM_008764266.1 | XP_008762488.1 | 12 - 13 | 7336 | 65 | ||
XM_008764267.1 | XP_008762489.1 | 13 - 14 | 2483 | 65 | ||
XM_008764268.1 | XP_008762490.1 | 11 - 12 | 2286 | 65 | ||
XM_008764269.1 | XP_008762491.1 | 12 - 13 | 2393 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |