Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009708.1 | NP_001009708.1 | 356 | 58 | ||
GenBank mRNA | BC087700.1 | - | 356 | 7314152 | 58 | |
FQ233103.1 | - | 199 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |