Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233347.2 | XP_006233409.1 | 1 - 2 | 170 | 112 | |
XM_008761534.1 | XP_008759756.1 | 1 - 2 | 170 | 112 | ||
XM_227694.7 | XP_227694.5 | 1 - 2 | 170 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |