Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015037.1 | NP_001015037.1 | 753 | 135 | ||
GenBank mRNA | AY955395.1 | - | 753 | 135 | ||
FQ218600.1 | - | 491 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |