Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168631.1 | NP_001162102.1 | 10 - 11 | 1879 | 73 | |
XM_006232079.2 | XP_006232141.1 | 10 - 11 | 1955 | 73 | ||
XM_006232080.2 | XP_006232142.1 | 10 - 11 | 1987 | 73 | ||
GenBank mRNA | AF468011.1 | - | 1 - 2 | 72 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |