Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138874.2 | NP_620229.2 | 25 - 26 | 752 | 78 | |
GenBank mRNA | J00710.1 | - | 25 - 26 | 752 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |