Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138874.2 | NP_620229.2 | 26 - 27 | 772 | 60 | |
GenBank mRNA | J00710.1 | - | 26 - 27 | 772 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |