Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191791.1 | NP_001178720.1 | 9 - 10 | 1012 | 62 | |
XM_006242719.2 | XP_006242781.1 | 9 - 10 | 1096 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |