Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sapcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004069.2 | NP_001004069.1 | 2 - 3 | 260 | 83 | |
XM_006256068.2 | XP_006256130.1 | 2 - 3 | 411 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |