Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181822.2 | NP_861543.2 | 1 - 2 | 221 | 60 | |
XM_006256054.2 | XP_006256116.1 | 1 - 2 | 391 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |