Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp21a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057101.2 | NP_476442.2 | 9 - 10 | 1211 | 137 | |
XM_006255907.2 | XP_006255969.1 | 9 - 10 | 1285 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |