Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748695.3 | XP_003748743.1 | 6 - 7 | 957 | 147 | |
XM_006227912.2 | XP_006227974.1 | 5 - 6 | 483 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |