Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039343.1 | NP_001034432.1 | 5 - 6 | 1625 | 73 | |
XM_006227884.2 | XP_006227946.1 | 5 - 6 | 1588 | 73 | ||
XM_006227885.2 | XP_006227947.1 | 5 - 6 | 1593 | 73 | ||
XM_006227886.2 | XP_006227948.1 | 5 - 6 | 1592 | 73 | ||
GenBank mRNA | AY886530.1 | - | 5 - 6 | 1625 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |