Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039343.1 | NP_001034432.1 | 7 - 8 | 1849 | 62 | |
XM_006227884.2 | XP_006227946.1 | 7 - 8 | 1812 | 62 | ||
XM_006227885.2 | XP_006227947.1 | 7 - 8 | 1817 | 62 | ||
XM_006227886.2 | XP_006227948.1 | 7 - 8 | 1816 | 62 | ||
GenBank mRNA | AY886530.1 | - | 7 - 8 | 1849 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus Probe spans exons |