Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Potec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751731.3 | XP_003751779.1 | 1414 | 72 | ||
XM_006254371.2 | XP_006254433.2 | 1 - 2 | 1414 | 72 | ||
XM_008771919.1 | XP_008770141.1 | 1414 | 72 | |||
XM_008771920.1 | XP_008770142.1 | 1414 | 72 | |||
XM_008771921.1 | XP_008770143.1 | 1414 | 72 | |||
GenBank mRNA | BC079322.1 | - | 580 | 7135443 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |