Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023093.1 | NP_075581.1 | 4 - 5 | 534 | 58 | |
GenBank mRNA | BC061830.1 | - | 4 - 5 | 534 | 5599635 | 58 |
FQ214031.1 | - | 3 - 4 | 376 | 58 | ||
FQ214146.1 | - | 3 - 4 | 376 | 58 | ||
J05266.1 | - | 4 - 5 | 455 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |