Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108728.1 | NP_001102198.1 | 9 - 10 | 1641 | 73 | |
XM_006240760.2 | XP_006240822.1 | 10 - 11 | 1546 | 73 | ||
XM_006240761.2 | XP_006240823.1 | 10 - 11 | 1558 | 73 | ||
XM_006240762.2 | XP_006240824.1 | 11 - 12 | 1666 | 73 | ||
XM_006240763.2 | XP_006240825.1 | 11 - 12 | 1654 | 73 | ||
XM_006240764.2 | XP_006240826.1 | 9 - 10 | 1450 | 73 | ||
XM_006240766.2 | XP_006240828.1 | 10 - 11 | 1761 | 73 | ||
XM_006240767.2 | XP_006240829.1 | 8 - 9 | 1210 | 73 | ||
XM_008765027.1 | XP_008763249.1 | 10 - 11 | 1580 | 73 | ||
GenBank mRNA | FQ220010.1 | - | 6 - 7 | 736 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |