Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244334.2 | XP_006244396.2 | 3 - 4 | 562 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |