Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_217313.8 | XP_217313.6 | 2 - 3 | 1971 | 98 | |
GenBank mRNA | FQ231323.1 | - | 2 - 3 | 258 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |