Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037660.1 | NP_001032749.1 | 11 - 12 | 1159 | 81 | |
GenBank mRNA | AJ628424.1 | - | 11 - 12 | 1159 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |