Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256704.2 | XP_006256766.1 | 1 - 2 | 271 | 68 | |
XM_006256705.2 | XP_006256767.1 | 1 - 2 | 272 | 68 | ||
GenBank mRNA | BC160871.1 | - | 1 - 2 | 46 | 7364965 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |