Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201420.2 | NP_958823.2 | 4 - 5 | 1022 | 113 | |
XM_006235691.2 | XP_006235753.1 | 4 - 5 | 991 | 113 | ||
GenBank mRNA | AY510602.1 | - | 4 - 5 | 813 | 113 | |
BC097260.1 | - | 4 - 5 | 1022 | 7443035 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |