Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133401.1 | NP_596892.1 | 7 - 8 | 763 | 77 | |
XM_006235994.2 | XP_006236056.1 | 9 - 10 | 1588 | 77 | ||
GenBank mRNA | AF257746.1 | - | 7 - 8 | 763 | 77 | |
AF286167.2 | - | 6 - 7 | 678 | 77 | ||
AY582535.1 | - | 6 - 7 | 678 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |