Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109486.1 | NP_001102956.1 | 5 - 6 | 398 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |