Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034939.1 | NP_001030111.2 | 1 - 2 | 479 | 71 | |
NM_001244819.1 | NP_001231748.1 | 2 - 3 | 202 | 71 | ||
XM_006240769.2 | XP_006240831.1 | 2 - 3 | 443 | 71 | ||
XM_006240770.2 | XP_006240832.1 | 2 - 3 | 298 | 71 | ||
XM_006240771.2 | XP_006240833.1 | 2 - 3 | 293 | 71 | ||
XM_008765029.1 | XP_008763251.1 | 2 - 3 | 458 | 71 | ||
GenBank mRNA | BC104710.1 | - | 1 - 2 | 479 | 7319804 | 71 |
DV214819.1 | - | 2 - 3 | 202 | 71 | ||
FQ219805.1 | - | 2 - 3 | 190 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |