Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017199.1 | NP_058895.1 | 1 - 2 | 220 | 137 | |
GenBank mRNA | BC058482.1 | - | 1 - 2 | 133 | 6921620 | 137 |
Z19087.1 | - | 1 - 2 | 220 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |