Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 9 - 10 | 984 | 95 | |
XM_006229603.2 | XP_006229665.1 | 8 - 9 | 1141 | 95 | ||
XM_006229604.2 | XP_006229666.1 | 8 - 9 | 3553 | 95 | ||
XM_006229605.2 | XP_006229667.1 | 9 - 10 | 2866 | 95 | ||
GenBank mRNA | BC169081.1 | - | 8 - 9 | 1048 | 5621009 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |