Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229562.2 | XP_006229624.1 | 2 - 3 | 858 | 131 | |
XM_006229563.2 | XP_006229625.1 | 2 - 3 | 709 | 131 | ||
XM_006229564.2 | XP_006229626.1 | 2 - 3 | 858 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Rattus norvegicus Probe spans exons |