Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004224.1 | NP_001004224.1 | 3 - 4 | 282 | 82 | |
XM_006229556.2 | XP_006229618.1 | 2 - 3 | 239 | 82 | ||
XM_006229557.2 | XP_006229619.1 | 3 - 4 | 356 | 82 | ||
GenBank mRNA | BC079182.1 | - | 3 - 4 | 282 | 7130880 | 82 |
FQ219566.1 | - | 3 - 4 | 273 | 82 | ||
FQ219790.1 | - | 3 - 4 | 281 | 82 | ||
FQ230132.1 | - | 3 - 4 | 273 | 82 | ||
FQ230136.1 | - | 3 - 4 | 273 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |