Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173434.1 | NP_001166905.1 | 3 - 4 | 744 | 96 | |
NM_001173435.1 | NP_001166906.1 | 2 - 3 | 444 | 96 | ||
NM_053301.4 | NP_445753.1 | 4 - 5 | 1008 | 96 | ||
GenBank mRNA | AF176534.1 | - | 3 - 4 | 664 | 96 | |
AF176535.1 | - | 2 - 3 | 364 | 96 | ||
AJ001517.1 | - | 4 - 5 | 1047 | 96 | ||
FQ219597.1 | - | 3 - 4 | 758 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |