Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053301.4 | NP_445753.1 | 1 - 2 | 169 | 114 | |
GenBank mRNA | AJ001517.1 | - | 1 - 2 | 208 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |