Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053301.4 | NP_445753.1 | 2 - 3 | 437 | 75 | |
GenBank mRNA | AF008587.1 | - | 1 - 2 | 236 | 75 | |
AJ001517.1 | - | 2 - 3 | 476 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |