Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034947.1 | NP_001030119.1 | 3 - 4 | 518 | 91 | |
GenBank mRNA | BC101920.1 | - | 3 - 4 | 518 | 7937874 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |