Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022960.2 | NP_075249.1 | 5 - 6 | 576 | 93 | |
XM_006243362.1 | XP_006243424.1 | 4 - 5 | 681 | 93 | ||
XM_008766336.1 | XP_008764558.1 | 1 - 2 | 382 | 93 | ||
GenBank mRNA | AF016406.1 | - | 1 - 2 | 355 | 93 | |
BC085731.1 | - | 5 - 6 | 576 | 7134715 | 93 | |
FQ209455.1 | - | 1 - 2 | 377 | 93 | ||
FQ211101.1 | - | 1 - 2 | 297 | 93 | ||
FQ212527.1 | - | 1 - 2 | 260 | 93 | ||
FQ214432.1 | - | 4 - 5 | 686 | 93 | ||
FQ218988.1 | - | 1 - 2 | 380 | 93 | ||
FQ219175.1 | - | 1 - 2 | 297 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |