Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106833.1 | NP_001100303.1 | 3 - 4 | 700 | 133 | |
XM_008766242.1 | XP_008764464.1 | 3 - 4 | 683 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |