Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213562.1 | NP_998727.1 | 1 - 2 | 110 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |