Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021764.1 | NP_068532.2 | 4 - 5 | 759 | 142 | |
XM_006235252.2 | XP_006235314.1 | 5 - 6 | 684 | 142 | ||
GenBank mRNA | AB011369.1 | - | 4 - 5 | 759 | 142 | |
BC061723.1 | - | 4 - 5 | 693 | 5598480 | 142 | |
U48248.1 | - | 4 - 5 | 759 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |