Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153624.2 | NP_705888.2 | 22 - 23 | 3576 | 72 | |
XM_006254403.2 | XP_006254465.1 | 22 - 23 | 3632 | 72 | ||
GenBank mRNA | BC166779.1 | - | 4 - 5 | 614 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |