Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079707.1 | NP_001073175.1 | 3 - 4 | 387 | 75 | |
XM_006242403.2 | XP_006242465.1 | 4 - 5 | 359 | 75 | ||
XM_008765750.1 | XP_008763972.1 | 4 - 5 | 349 | 75 | ||
XM_008765751.1 | XP_008763973.1 | 4 - 5 | 351 | 75 | ||
XM_008765752.1 | XP_008763974.1 | 4 - 5 | 420 | 75 | ||
GenBank mRNA | BC128778.1 | - | 3 - 4 | 387 | 7134533 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |