Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030998.1 | NP_112260.1 | 4 - 5 | 578 | 64 | |
GenBank mRNA | KJ923229.1 | - | 4 - 5 | 578 | 64 | |
U42427.1 | - | 4 - 5 | 578 | 64 | ||
X71916.1 | - | 4 - 5 | 560 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |