Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013106.1 | NP_001013124.1 | 10 - 11 | 1092 | 63 | |
GenBank mRNA | BC091366.1 | - | 10 - 11 | 1092 | 7319183 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |