Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106794.2 | NP_001100264.2 | 2 - 3 | 223 | 119 | |
GenBank mRNA | BC169128.1 | - | 2 - 3 | 189 | 6921287 | 119 |
CF976389.1 | - | 2 - 3 | 227 | 119 | ||
CF976393.1 | - | 2 - 3 | 194 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |