Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vangl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109584.1 | NP_001103054.1 | 1 - 2 | 99 | 68 | |
XM_006233054.2 | XP_006233116.1 | 1 - 2 | 140 | 68 | ||
XM_006233055.2 | XP_006233117.1 | 1 - 2 | 139 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |