Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bnipl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128187.2 | NP_001121659.1 | 8 - 9 | 977 | 81 | |
XM_006232889.2 | XP_006232951.1 | 7 - 8 | 1316 | 81 | ||
GenBank mRNA | BC166747.1 | - | 8 - 9 | 977 | 5598850 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |