Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107709.1 | NP_001101179.1 | 6 - 7 | 1599 | 74 | |
XM_006233081.2 | XP_006233143.1 | 7 - 8 | 1466 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |